Masterflex® Printed Microscope Slide, ten rectangular sections; 144/pk
# de catalogue MFLX75955-31
Fournisseur: VWR International
Spécifications
- Description:Microscope slides, printed with mask, ten rectangular sections
- L×W:76.2×25.4 mm (3×1")
- Cat. no.:MFLX75955-31
- Slide Type:Printed without Mask
Spécifications
A propos de cet article
Optimize sample testing and viewing.
- Screen-printed section design keeps samples contained
- Hydrophobic epoxy ink withstands frequent washing and resists chemicals
- Contrasting ink makes orienting the microscope field of view over a sample easy
Printed microscope slides are available with a variety of section shapes, sizes, counts, colors, and labels. The PTFE-based epoxy ink creates a hydrophobic barrier, holding samples in place for easy viewing under a microscope, and keeping reactions isolated from each other in multi-section slides.
- Description:Microscope slides, printed with mask, ten rectangular sections
- L×W:76.2×25.4 mm (3×1")
- Cat. no.:MFLX75955-31
- Slide Type:Printed without Mask
Customers Who Bought This Also Bought