Masterflex® Printed Agglutination Slide, 12 circles, not numbered; 144/pk
# de catalogue MFLX75955-05
Fournisseur: VWR International
Spécifications
- Description:Printed agglutination slides, not numbered
- L×W:76.2×50.8 mm (3×2")
- Material:Glass
- Cat. no.:MFLX75955-05
- Well Shape:Round
Spécifications
A propos de cet article
No more makeshift slides.
- Large wells clearly show reaction results
- Hydrophobic epoxy ink withstands frequent washing and resists chemicals
- Choose numbered/unnumbered—Unnumbered slides allow you to customize the labeling
Use these printed slides to perform latex and other agglutination tests for quick microbe identification. Choose between eight or twelve wells, square or circular, and numbered or unnumbered to suit your application.
- Description:Printed agglutination slides, not numbered
- L×W:76.2×50.8 mm (3×2")
- Material:Glass
- Cat. no.:MFLX75955-05
- Well Shape:Round
Customers Who Bought This Also Bought