Masterflex® Sanitary Fitting Clamps, PVDF, Avantor®
Supplier: VWR International
Certificates
About this item
Maintain sanitary conditions in your processes.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These sanitary clamps are compatible with Tri-Clamp® fittings. The PVDF material has excellent chemical resistance with low leachable characteristics.
Specifications
- Catalog No:
- MFLX31811-02
- MFLX31811-01
- MFLX31811-00
- Model:
- 31811-02
- 31811-01
- 31811-00
- Sanitary clamp size (port 1):
- 2"
- 1-1/2"
- 3/4"
- Connection type:
- Sanitary
- Sanitary
- Sanitary
- Chemical resistance:
- Very good
- Very good
- Very good
- Min. temperature:
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- Max. temperature:
- 138 °C (280 °F)
- 138 °C (280 °F)
- 138 °C (280 °F)
- Max. operating pressure:
- 150 psi
- 150 psi
- 150 psi
- Sterilisation:
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Warranty:
- 1 year
- 1 year
- 1 year