Masterflex® Sanitary Clamp Fittings (Nylon), Avantor®
Fournisseur: VWR International
Certificats
A propos de cet article
Compatible with metal and plastic Tri-Clamp® fittings.
- Excellent chemical resistance; low leachable characteristics
- Autoclavable
Constructed of nylon, these Tri-Clamp® sanitary clamps are perfect for single-use transfer lines in biopharma markets, plus a wide range of fluid transfer systems. These clamps ideal for securing your fluid transfer system without damaging the process lines.
Spécifications
- Model:
- 30508-25
- 30508-27
- 30508-21
- 30508-23
- 30508-29
- Sterilisation:
- Autoclave or Ethylene oxide (ETO)
- Autoclave or Ethylene oxide (ETO)
- Autoclave or Ethylene oxide (ETO)
- Ethylene oxide (ETO) and autoclavable
- Autoclave or Ethylene oxide (ETO)
- Warranty:
- Manufacturer defect
- Manufacturer defect
- Manufacturer defect
- Manufacturer defect
- Manufacturer defect