Masterflex® Turbine Flowmeter with Ultra-High Accuracy, Ryton Housing; 100 mL/min
# de catalogue MFLX32714-00
Fournisseur: MC MILLAN CO
CE Compliant
NIST traceable
Spécifications
- Accuracy:±0.5% full-scale
- Display type:3.5-digit LCD. 0.875"H
- Maximum Flow Rate:100 ml/min
- Min. flow rate:13 ml/min
- Output:0 to 5 VDC
- Description:High-Accuracy Turbine Meter, Ryton, 13 to 100 ml/min
- Max. operating temperature:50 °C (122 °F)
- Height:15862"
- Repeatability:±0.2% full-scale
- Warranty:1 year
- Cat. no.:MFLX32714-00
- Housing Material:Ryton
Spécifications
A propos de cet article
Provides ±0.5% full scale accuracy in a compact, in-line liquid flowmeter.
- Test 02a_BulletPoint
- Withstands up to 500 psi
- Simultaneously displays flow rate
Test 03a_BodyText
Select from economical Ryton® construction or rugged brass to withstand up to 500 psi. Flowmeters simultaneously display flow rate, plus provide a signal output for datalogging or control. Power your flowmeters with optional AC adapter kits with signal cables, sold separately.
Test 04a_BodyText
Delivery: Test 11a_Delivery
Delivery: Includes nIST-traceable calibration certificate.
- Accuracy:±0.5% full-scale
- Display type:3.5-digit LCD. 0.875"H
- Maximum Flow Rate:100 ml/min
- Min. flow rate:13 ml/min
- Output:0 to 5 VDC
- Description:High-Accuracy Turbine Meter, Ryton, 13 to 100 ml/min
- Max. operating temperature:50 °C (122 °F)
- Height:15862"
- Repeatability:±0.2% full-scale
- Warranty:1 year
- Cat. no.:MFLX32714-00
- Housing Material:Ryton
Customers Who Bought This Also Bought