Masterflex® Correlated Flowmeters with High-Res Valves, 150-mm Scale, Avantor®
Supplier: Avantor
Certificates
About this item
Don’t sacrifice suitability for precision.
- Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
Specifications
- Catalog No:
- MFLX03217-74
- MFLX03217-76
- MFLX03227-08
- MFLX03227-04
- MFLX03227-12
- MFLX03229-31
- MFLX03217-79
- MFLX03295-14
- MFLX03295-22
- MFLX03295-20
- MFLX03229-27
- MFLX03295-26
- MFLX03229-29
- MFLX03295-24
- MFLX03295-28
- MFLX03229-17
- MFLX03229-19
- MFLX03229-21
- MFLX03217-58
- MFLX03229-23
- MFLX03229-07
- MFLX03217-63
- MFLX03229-11
- MFLX03217-68
- MFLX03229-15
- MFLX03295-06
- MFLX03295-04
- MFLX03295-12
- MFLX03295-08
- MFLX03217-72
- MFLX03227-28
- MFLX03227-30
- MFLX03217-73
- MFLX03227-18
- MFLX03227-22
- MFLX03227-26
- MFLX03227-20
- MFLX03227-14
- MFLX03227-84
- MFLX03227-24
- MFLX03217-67
- MFLX03229-09
- MFLX03227-06
- MFLX03229-05
- MFLX03227-10
- MFLX03229-25
- MFLX03227-16
- MFLX03229-13
- MFLX03217-57
- MFLX03295-18
- MFLX03217-62
- MFLX03217-53
- MFLX03295-10
- MFLX03295-16
- Model:
- 03217-74
- 03217-76
- 03227-08
- 03227-04
- 03227-12
- 03229-31
- 03217-79
- 03295-14
- 03295-22
- 03295-20
- 03229-27
- 03295-26
- 03229-29
- 03295-24
- 03295-28
- 03229-17
- 03229-19
- 03229-21
- 03217-58
- 03229-23
- 03229-07
- 03217-63
- 03229-11
- 03217-68
- 03229-15
- 03295-06
- 03295-04
- 03295-12
- 03295-08
- 03217-72
- 03227-28
- 03227-30
- 03217-73
- 03227-18
- 03227-22
- 03227-26
- 03227-20
- 03227-14
- 03227-84
- 03227-24
- 03217-67
- 03229-09
- 03227-06
- 03229-05
- 03227-10
- 03229-25
- 03227-16
- 03229-13
- 03217-57
- 03295-18
- 03217-62
- 03217-53
- 03295-10
- 03295-16
- Accuracy:
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- Float material:
- Sapphire
- Sapphire
- Glass
- Glass
- Glass
- 316 Stainless Steel
- Glass
- 316 Stainless Steel
- 316 Stainless Steel
- Glass
- 316 Stainless Steel
- 316 Stainless Steel
- Glass
- Glass
- Glass
- Glass
- 316 Stainless Steel
- Glass
- Glass
- 316 Stainless Steel
- 316 Stainless Steel
- Glass
- 316 Stainless Steel
- Glass
- 316 Stainless Steel
- 316 Stainless Steel
- Glass
- Glass
- Glass
- Sapphire
- Glass
- 316 Stainless Steel
- Sapphire
- 316 Stainless Steel
- 316 Stainless Steel
- 316 Stainless Steel
- Glass
- 316 Stainless Steel
- Glass
- Glass
- Sapphire
- Glass
- 316 Stainless Steel
- Glass
- 316 Stainless Steel
- Glass
- Glass
- Glass
- Sapphire
- 316 Stainless Steel
- Sapphire
- Glass
- 316 Stainless Steel
- Glass
- Flow Tube material:
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Max. air flow rate:
- 5005 ml/min
- 11356 ml/min
- 92 ml/min
- 49.1 ml/min
- 374 ml/min
- 16737 ml/min
- 2313 ml/min
- 814 ml/min
- 4562 ml/min
- 2313 ml/min
- 7825 ml/min
- 7825 ml/min
- 8678 ml/min
- 3807 ml/min
- 8678 ml/min
- 844 ml/min
- 1682 ml/min
- 2313 ml/min
- 92 ml/min
- 4562 ml/min
- 137 ml/min
- 374 ml/min
- 264 ml/min
- 825 ml/min
- 814 ml/min
- 137 ml/min
- 49.1 ml/min
- 374 ml/min
- 92 ml/min
- 1093 ml/min
- 8678 ml/min
- 16737 ml/min
- 3079 ml/min
- 1682 ml/min
- 4562 ml/min
- 7825 ml/min
- 2313 ml/min
- 814 ml/min
- 8678 ml/min
- 3807 ml/min
- 513 ml/min
- 92 ml/min
- 137 ml/min
- 49.1 ml/min
- 264 ml/min
- 3807 ml/min
- 844 ml/min
- 374 ml/min
- 73 ml/min
- 1682 ml/min
- 140 ml/min
- 49 ml/min
- 264 ml/min
- 844 ml/min
- Max. argon flow rate:
- 4388 ml/min
- 9557 ml/min
- 75.6 ml/min
- 53.5 ml/min
- 305 ml/min
- 14131 ml/min
- 2048 ml/min
- 676 ml/min
- 3903 ml/min
- 1949 ml/min
- 6384 ml/min
- 6384 ml/min
- 7366 ml/min
- 3151 ml/min
- 7366 ml/min
- 687 ml/min
- 1380 ml/min
- 1949 ml/min
- 103.1 ml/min
- 3903 ml/min
- 113 ml/min
- 355 ml/min
- 218 ml/min
- 725 ml/min
- 676 ml/min
- 113 ml/min
- 53.5 ml/min
- 305 ml/min
- 75.6 ml/min
- 944 ml/min
- 7366 ml/min
- 14131 ml/min
- 2620 ml/min
- 1380 ml/min
- 3903 ml/min
- 6384 ml/min
- 1949 ml/min
- 676 ml/min
- 7366 ml/min
- 3151 ml/min
- 472 ml/min
- 75.6 ml/min
- 113 ml/min
- 53.5 ml/min
- 218 ml/min
- 3151 ml/min
- 687 ml/min
- 305 ml/min
- 84 ml/min
- 1380 ml/min
- 157 ml/min
- 56.8 ml/min
- 218 ml/min
- 687 ml/min
- Max. CO₂ flow rate:
- 4175 ml/min
- 9539 ml/min
- 103.1 ml/min
- 56.4 ml/min
- 355 ml/min
- 14051 ml/min
- 1949 ml/min
- 728 ml/min
- 3990 ml/min
- 2048 ml/min
- 6308 ml/min
- 6308 ml/min
- 7485 ml/min
- 3374 ml/min
- 7485 ml/min
- 725 ml/min
- 1420 ml/min
- 2048 ml/min
- 75.6 ml/min
- 3990 ml/min
- 150 ml/min
- 305 ml/min
- 281 ml/min
- 687 ml/min
- 728 ml/min
- 150 ml/min
- 56.4 ml/min
- 355 ml/min
- 103.1 ml/min
- 910 ml/min
- 7485 ml/min
- 14051 ml/min
- 2605 ml/min
- 1420 ml/min
- 3990 ml/min
- 6308 ml/min
- 2048 ml/min
- 728 ml/min
- 7485 ml/min
- 3374 ml/min
- 429 ml/min
- 103.1 ml/min
- 150 ml/min
- 56.4 ml/min
- 281 ml/min
- 3374 ml/min
- 725 ml/min
- 355 ml/min
- 60.1 ml/min
- 1420 ml/min
- 110.9 ml/min
- 43.5 ml/min
- 281 ml/min
- 725 ml/min
- Max. N₂ flow rate:
- 5090 ml/min
- 11524 ml/min
- 92 ml/min
- 48 ml/min
- 382 ml/min
- 17021 ml/min
- 2395 ml/min
- 824 ml/min
- 4685 ml/min
- 2395 ml/min
- 7722 ml/min
- 7722 ml/min
- 8916 ml/min
- 3868 ml/min
- 8916 ml/min
- 827 ml/min
- 1662 ml/min
- 2395 ml/min
- 92 ml/min
- 4685 ml/min
- 143 ml/min
- 382 ml/min
- 271 ml/min
- 827 ml/min
- 824 ml/min
- 143 ml/min
- 48 ml/min
- 382 ml/min
- 92 ml/min
- 1110 ml/min
- 8916 ml/min
- 17021 ml/min
- 3142 ml/min
- 1662 ml/min
- 4685 ml/min
- 7722 ml/min
- 2395 ml/min
- 824 ml/min
- 8916 ml/min
- 3868 ml/min
- 520 ml/min
- 92 ml/min
- 143 ml/min
- 48 ml/min
- 271 ml/min
- 3868 ml/min
- 827 ml/min
- 382 ml/min
- 76 ml/min
- 1662 ml/min
- 139 ml/min
- 56 ml/min
- 271 ml/min
- 827 ml/min
- Max. helium flow rate:
- 10336 ml/min
- 25400 ml/min
- 90.1 ml/min
- 46.2 ml/min
- 450 ml/min
- 38576 ml/min
- 4880 ml/min
- 1290 ml/min
- 9770 ml/min
- 4880 ml/min
- 15960 ml/min
- 15960 ml/min
- 19426 ml/min
- 7803 ml/min
- 19426 ml/min
- 1490 ml/min
- 3397 ml/min
- 4880 ml/min
- 90.1 ml/min
- 9770 ml/min
- 133 ml/min
- 450 ml/min
- 283 ml/min
- 1490 ml/min
- 1290 ml/min
- 133 ml/min
- 46.2 ml/min
- 450 ml/min
- 90.1 ml/min
- 2059 ml/min
- 19426 ml/min
- 38576 ml/min
- 6458 ml/min
- 3397 ml/min
- 9770 ml/min
- 15960 ml/min
- 4880 ml/min
- 1290 ml/min
- 19426 ml/min
- 7803 ml/min
- 681 ml/min
- 90.1 ml/min
- 133 ml/min
- 46.2 ml/min
- 283 ml/min
- 7803 ml/min
- 1490 ml/min
- 450 ml/min
- 69 ml/min
- 3397 ml/min
- 142 ml/min
- 47 ml/min
- 283 ml/min
- 1490 ml/min
- Max. H₂ flow rate:
- 16108 ml/min
- 40006 ml/min
- 208 ml/min
- 94 ml/min
- 1027 ml/min
- 59996 ml/min
- 7817 ml/min
- 2496 ml/min
- 15855 ml/min
- 7817 ml/min
- 27804 ml/min
- 27804 ml/min
- 2940 ml/min
- 13105 ml/min
- 2940 ml/min
- 2620 ml/min
- 5547 ml/min
- 7817 ml/min
- 208 ml/min
- 15855 ml/min
- 301 ml/min
- 1021 ml/min
- 627 ml/min
- 2620 ml/min
- 2496 ml/min
- 301 ml/min
- 94 ml/min
- 1027 ml/min
- 208 ml/min
- 3546 ml/min
- 2940 ml/min
- 59996 ml/min
- 10455 ml/min
- 5547 ml/min
- 15855 ml/min
- 27804 ml/min
- 7817 ml/min
- 2496 ml/min
- 29840 ml/min
- 13105 ml/min
- 1497 ml/min
- 208 ml/min
- 301 ml/min
- 94 ml/min
- 627 ml/min
- 13105 ml/min
- 2620 ml/min
- 1027 ml/min
- 149 ml/min
- 5547 ml/min
- 322 ml/min
- 100 ml/min
- 627 ml/min
- 2620 ml/min
- Max. O₂ flow rate:
- 4652 ml/min
- 10706 ml/min
- 81 ml/min
- 42 ml/min
- 340 ml/min
- 15710 ml/min
- 2169 ml/min
- 753 ml/min
- 4341 ml/min
- 2169 ml/min
- 6992 ml/min
- 6992 ml/min
- 8269 ml/min
- 3485 ml/min
- 8269 ml/min
- 772 ml/min
- 1545 ml/min
- 2169 ml/min
- 81 ml/min
- 4341 ml/min
- 131 ml/min
- 340 ml/min
- 233 ml/min
- 772 ml/min
- 753 ml/min
- 131 ml/min
- 42 ml/min
- 340 ml/min
- 81 ml/min
- 1024 ml/min
- 8269 ml/min
- 15710 ml/min
- 2860 ml/min
- 1545 ml/min
- 4341 ml/min
- 6992 ml/min
- 2169 ml/min
- 753 ml/min
- 8269 ml/min
- 3485 ml/min
- 472 ml/min
- 81 ml/min
- 131 ml/min
- 42 ml/min
- 233 ml/min
- 3485 ml/min
- 772 ml/min
- 340 ml/min
- 70 ml/min
- 1545 ml/min
- 121 ml/min
- 49 ml/min
- 233 ml/min
- 772 ml/min
- Max. operating temperature:
- 65 °C (150 °F)
- 65 °C (150 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 65 °C (150 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 65 °C (150 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 65 °C (150 °F)
- 121 °C (250 °F)
- 65 °C (150 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 65 °C (150 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 65 °C (150 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 65 °C (150 °F)
- 121 °C (250 °F)
- 65 °C (150 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 65 °C (150 °F)
- 121 °C (250 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- Max. water flow rate:
- 126 ml/min
- 306 ml/min
- 0.85 ml/min
- 0.49 ml/min
- 5.5 ml/min
- 506 ml/min
- 53.5 ml/min
- 20.4 ml/min
- 134 ml/min
- 53.5 ml/min
- 217 ml/min
- 217 ml/min
- 210 ml/min
- 84 ml/min
- 210 ml/min
- 16.5 ml/min
- 46 ml/min
- 53.5 ml/min
- 0.85 ml/min
- 134 ml/min
- 2.45 ml/min
- 5.5 ml/min
- 4.74 ml/min
- 16.5 ml/min
- 20.4 ml/min
- 2.45 ml/min
- 0.49 ml/min
- 5.5 ml/min
- 0.85 ml/min
- 26.1 ml/min
- 210 ml/min
- 506 ml/min
- 77.8 ml/min
- 46 ml/min
- 134 ml/min
- 217 ml/min
- 53.5 ml/min
- 20.4 ml/min
- 210 ml/min
- 84 ml/min
- 9.9 ml/min
- 0.85 ml/min
- 2.45 ml/min
- 0.49 ml/min
- 4.74 ml/min
- 84 ml/min
- 16.5 ml/min
- 5.5 ml/min
- 0.99 ml/min
- 46 ml/min
- 1.92 ml/min
- 0.53 ml/min
- 4.74 ml/min
- 16.5 ml/min
- Max. pressure:
- 100 psi
- 100 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 100 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 100 psi
- 200 psi
- 200 psi
- 100 psi
- 200 psi
- 100 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 100 psi
- 200 psi
- 200 psi
- 100 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 100 psi
- 200 psi
- 100 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 100 psi
- 200 psi
- 100 psi
- 100 psi
- 200 psi
- 200 psi
- O-Ring material:
- PTFE
- PTFE
- Buna-N
- Buna-N
- Buna-N
- Viton®
- PTFE
- Buna-N
- Buna-N
- Buna-N
- Viton®
- Buna-N
- Viton®
- Buna-N
- Buna-N
- Viton®
- Viton®
- Viton®
- PTFE
- Viton®
- Viton®
- PTFE
- Viton®
- PTFE
- Viton®
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- PTFE
- Buna-N
- Buna-N
- PTFE
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- PTFE
- Buna-N
- PTFE
- Viton®
- Buna-N
- Viton®
- Buna-N
- Viton®
- Buna-N
- Viton®
- PTFE
- Buna-N
- PTFE
- PTFE
- Buna-N
- Buna-N
- Repeatability:
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year