Masterflex® Correlated Variable-Area Flowmeters with Valve, 150-mm Scale, Avantor®
Fournisseur: Avantor
Certificats
A propos de cet article
Don’t sacrifice suitability for precision.
- Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
Spécifications
- Catalog No:
- MFLX03217-70
- MFLX03217-75
- MFLX03294-14
- MFLX03294-24
- MFLX03294-26
- MFLX03217-77
- MFLX03294-06
- MFLX03294-08
- MFLX03217-80
- MFLX03294-10
- MFLX03294-12
- MFLX03217-24
- MFLX03217-26
- MFLX03294-04
- MFLX03217-28
- MFLX03217-20
- MFLX03217-14
- MFLX03219-27
- MFLX03217-22
- MFLX03219-17
- MFLX03219-19
- MFLX03219-21
- MFLX03219-23
- MFLX03219-11
- MFLX03219-13
- MFLX03219-15
- MFLX03217-16
- MFLX03217-18
- MFLX03217-30
- MFLX03217-65
- MFLX03219-25
- MFLX03219-29
- MFLX03217-12
- MFLX03219-31
- MFLX03217-66
- MFLX03294-18
- MFLX03294-16
- MFLX03294-30
- MFLX03217-06
- MFLX03217-71
- MFLX03217-08
- MFLX03217-10
- MFLX03217-04
- MFLX03217-85
- MFLX03219-05
- MFLX03217-81
- MFLX03219-07
- MFLX03219-09
- MFLX03217-51
- MFLX03217-60
- MFLX03217-61
- MFLX03217-55
- MFLX03217-56
- Model:
- 03217-70
- 03217-75
- 03294-14
- 03294-24
- 03294-26
- 03217-77
- 03294-06
- 03294-08
- 03217-80
- 03294-10
- 03294-12
- 03217-24
- 03217-26
- 03294-04
- 03217-28
- 03217-20
- 03217-14
- 03219-27
- 03217-22
- 03219-17
- 03219-19
- 03219-21
- 03219-23
- 03219-11
- 03219-13
- 03219-15
- 03217-16
- 03217-18
- 03217-30
- 03217-65
- 03219-25
- 03219-29
- 03217-12
- 03219-31
- 03217-66
- 03294-18
- 03294-16
- 03294-30
- 03217-06
- 03217-71
- 03217-08
- 03217-10
- 03217-04
- 03217-85
- 03219-05
- 03217-81
- 03219-07
- 03219-09
- 03217-51
- 03217-60
- 03217-61
- 03217-55
- 03217-56
- Float material:
- Sapphire
- Sapphire
- 316 SS
- Glass
- 316 SS
- 316 SS
- Glass
- Sapphire
- 316 SS
- Glass
- Glass
- 316 SS
- Glass
- Glass
- Glass
- 316 SS
- 316 SS
- 316 SS
- Glass
- 316 SS
- Glass
- 316 SS
- 316 SS
- Glass
- 316 SS
- Glass
- 316 SS
- 316 SS
- Sapphire
- Glass
- Glass
- Glass
- 316 SS
- 316 SS
- Glass
- 316 SS
- 316 SS
- Glass
- 316 SS
- Glass
- Sapphire
- Glass
- 316 SS
- Glass
- Sapphire
- Sapphire
- Glass
- Flow Tube material:
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Max. air flow rate:
- 1093 ml/min
- 3079 ml/min
- 814 ml/min
- 3807 ml/min
- 7825 ml/min
- 3807 ml/min
- 137 ml/min
- 92 ml/min
- 5005 ml/min
- 264 ml/min
- 374 ml/min
- 3807 ml/min
- 7825 ml/min
- 49.1 ml/min
- 8678 ml/min
- 2313 ml/min
- 814 ml/min
- 7825 ml/min
- 4562 ml/min
- 844 ml/min
- 1682 ml/min
- 2313 ml/min
- 4562 ml/min
- 264 ml/min
- 374 ml/min
- 814 ml/min
- 844 ml/min
- 1682 ml/min
- 16737 ml/min
- 513 ml/min
- 3807 ml/min
- 8678 ml/min
- 374 ml/min
- 16737 ml/min
- 825 ml/min
- 1682 ml/min
- 844 ml/min
- 16737 ml/min
- 137 ml/min
- 2313 ml/min
- 92 ml/min
- 264 ml/min
- 49.1 ml/min
- 11356 ml/min
- 49.1 ml/min
- 8678 ml/min
- 137 ml/min
- 92 ml/min
- 49 ml/min
- 140 ml/min
- 374 ml/min
- 73 ml/min
- 92 ml/min
- Max. operating temperature:
- 65 °C (150 °F)
- 65 °C (150 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 65 °C (150 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 65 °C (150 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 65 °C (150 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 65 °C (150 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 65 °C (150 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 65 °C (150 °F)
- 121 °C (250 °F)
- 65 °C (150 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- O-Ring material:
- PTFE
- PTFE
- Buna-N
- Buna-N
- Buna-N
- PTFE
- Buna-N
- Buna-N
- PTFE
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Viton®
- Buna-N
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Buna-N
- Buna-N
- Buna-N
- PTFE
- Viton®
- Viton®
- Buna-N
- Viton®
- PTFE
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- PTFE
- Buna-N
- Buna-N
- Buna-N
- PTFE
- Viton®
- PTFE
- Viton®
- Viton®
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- Repeatability:
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25% full-scale
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25% full-scale
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25% full-scale
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25% full-scale
- ±0.25%
- ±0.25%
- ±0.25% full-scale
- ±0.25%
- ±0.25% full-scale
- ±0.25%
- ±0.25% full-scale
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year