Masterflex® L/S® Easy-Load® Pump Heads for Precision Tubing, Avantor®
Fournisseur: Avantor Fluid Handling
Certificats
A propos de cet article
Save time with fast tubing changes and reduced maintenance.
- New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
- Rotor is constructed of stainless steel (SS) for enhanced durability
- Pump heads are stackable for multichannel applications
- Compact design for efficient use of space
- Suitable for use with L/S® Precision pump tubing sizes
This Easy-Load® pump head accepts precision tubing— ensures optimal performance from your Masterflex pump. Select a pump head for one of the following precision tubing sizes: L/S 13, L/S 14, L/S 16, L/S 25, L/S 17, L/S 18.
Spécifications
- Catalog No:
- MFLX07514-40
- MFLX07514-30
- MFLX07514-10
- Model:
- 07514-40
- 07514-30
- 07514-10
- Gear ratio:
- 1:1
- 1:1
- 1:1
- No. of channels:
- 1
- 1
- 1
- Occlusion:
- Fixed
- Fixed
- Fixed
- Open-head sensor:
- No
- Yes
- No
- Optimum RPM range:
- 0.02 - 600 rpm
- 0.02 - 600 rpm
- 0.02 - 600 rpm
- Pump series:
- L/S
- L/S
- L/S