Masterflex® Dual-Drive Syringe Pump Accessories, Avantor®
Fournisseur: VWR International
Certificats
A propos de cet article
Daisy chain up to 100 pumps.
- Connect multiple independent syringes to operate simultaneously
- Easily connect pumps
- Choose cable based on your current pump
Optimize performance of your Masterflex® Dual-Drive Syringe Pump with these accessories.
Spécifications
- Catalog No:
- MFLX74901-87
- MFLX74901-88
- Model:
- 74901-87
- 74901-88
- Outlet connection:
- Daisy Chain Cable for 74901-80
- RS-232 Connector Cable for 74901-80
- Warranty:
- 1 year
- 1 year