Masterflex® Six-Channel Syringe Pumps, Avantor®
Fournisseur: VWR International
Certificats
A propos de cet article
Six-channel pump can both withdraw and infuse.
- Flow rates up to 22.3 ml/min per syringe
- Network up to 100 pumps from a single communications port on a computer
- Economical pump can both withdraw and infuse
- Pumps are capable of holding 41 commands, do unlimited repetitions and unlimited runtime
- Built-i
Programming commands allow dispensing of a specified amount, automatic rates changes, direction changes, programmed pauses, external or user input, and sending or receive signals from other equipment. Non-volatile memory restores all setup parameters and
Spécifications
- Catalog No:
- MFLX75900-50
- MFLX75900-55
- Accuracy:
- ±1%
- ±1%
- Linear force:
- 100 lbs.
- 100 lbs.
- Reproducibility:
- ±1%
- ±1%
- Syringe size accepted:
- 60 ml
- 60 ml
- Warranty:
- 2 years
- 2 years