Masterflex® Micro Peristaltic Pumps, Avantor®
Fournisseur: VWR International
Certificats
A propos de cet article
Operate by computer or on your iOS or Android™ device.
- Easily achieve flow ranges down to 0.2 µL/min with a 1.26% reproducibility
- 12-roller head minimizes the pulsation
- Select pump based upon number of channels needed—from one to eight
- Wide choice of peristaltic tube sets (0.13 mm i.d. to 3.17 mm i.d.)
Control these micro pumps through your computer using an available USB port and Pump Network Control™ software. Software† can be used remotely on the same Wi-Fi network using an Apple® iOS or Android device. The Pump Network Control software controls dire
Spécifications
- Min. flow rate:
- 0.2 µl/min
- 0.2 µl/min
- 0.2 µl/min
- 0.2 µl/min
- CE regulatory approval:
- Yes
- Yes
- Yes
- Yes
- Self-priming:
- Yes
- Yes
- Yes
- Yes
- Shipping weight:
- 2.15 kg (4.75 lbs.)
- 2.15 kg (4.75 lbs.)
- 2.15 kg (4.75 lbs.)
- 2.15 kg (4.75 lbs.)
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year