Masterflex® Stepping Motor Proportioning Valves, Avantor®
Fournisseur: AALBORG INSTRUMENTS & CONTROL
Certificats
A propos de cet article
Resist pressures up to 500 psi!
- High precision, two-way metering valves in aluminum or 316 SS for air/water
- Operate continuously without coil overheating problems found in traditional solenoid designs
Stainless steel models include a 316 SS valve blocks, PTFE-lined aluminum housing blocks, Viton® O-rings, and PFA closing pins.
Aluminum models include aluminum housings and valve blocks, Viton® O-rings, and PFA closing pins.
Also includes a 9-pin "D" electrical connector. Optional connection cable (98651-50) for easy input/output wiring. Power input is +12 VDC at 800 mA, protected by a resettable fuse.
Spécifications
- Model:
- 98651-00
- 98651-06
- 98651-08
- 98651-10
- 98651-04
- 98651-02
- Height:
- 6.4"
- 6.8"
- 6.8"
- 6.8"
- 6.8"
- 6.4"
- Max. pressure:
- 500 psi
- 500 psi
- 500 psi
- 500 psi
- 500 psi
- 500 psi
- Power:
- 12 VDC
- 12 VDC
- 12 VDC
- 12 VDC
- 12 VDC
- 12 VDC
- Tubing ext.Ø:
- 0.375"
- 0.5"
- 0.5"
- 0.375"
- Wetted materials:
- Aluminum, Viton, PFA
- SS, Viton, PFA
- Aluminum, Viton, PFA
- SS, Viton, PFA
- Aluminum, Viton, PFA
- SS, Viton, PFA
- Width:
- 5.2"
- 5.4"
- 3"
- 3"
- 5.4"
- 5.2"