Masterflex® Inert In-Line Check Valves, PEEK, Avantor®
Fournisseur: VWR International
Certificats
A propos de cet article
No maintenance required!
- Ideal for high-purity, low pressure
- No metal components — inert flow path
- No back flow leakage with back pressure of 0 to 100 psi
- 1/4-28 UNF threaded connections
Unlike spring-actuated check valves that may restrict or impede the flow path, this in-line check valve features a smooth flow path that minimizes shear and turbulence.
Spécifications
- Model:
- 01355-24
- 01355-28
- 01355-04
- 01355-12
- 01355-16
- 01355-00
- Max. pressure:
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- Wetted materials:
- PEEK/EPDM
- PEEK
- PEEK
- PEEK/EPDM
- PEEK
- PEEK/EPDM
- Warranty:
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years