Masterflex® Transfer Tubing, PharMed® BPT, Avantor®
Fournisseur: VWR International
Certificats
A propos de cet article
Protect your light-sensitive samples.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. FDA compliant and made according to GMP. Temperature range is -51 to 270 °F (-51 to 132 °C). Can be sterilized with ethylene oxide, gamma, or autoclave.
Spécifications
- Catalog No:
- MFLX96880-18
- MFLX96880-16
- MFLX96880-15
- MFLX96880-12
- MFLX96880-03
- MFLX96880-02
- MFLX96880-30
- MFLX96880-22
- MFLX96880-00
- MFLX96880-20
- MFLX96880-06
- MFLX96880-04
- MFLX96880-14
- MFLX96880-10
- MFLX96880-05
- MFLX96880-08
- Model:
- 96880-18
- 96880-16
- 96880-15
- 96880-12
- 96880-03
- 96880-02
- 96880-30
- 96880-22
- 96880-00
- 96880-20
- 96880-06
- 96880-04
- 96880-14
- 96880-10
- 96880-05
- 96880-08
- GMP compliant:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Length per pack:
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- 7.6 m (25 ft.)
- Max. pressure:
- 0.7584236 bar (11 psi)
- 0.8963188 bar (13 psi)
- 0.5515808 bar (8 psi)
- 1.3100044 bar (19 psi)
- 2.757904 bar (40 psi)
- 2.757904 bar (40 psi)
- 0.8963188 bar (13 psi)
- 0.4826332 bar (7 psi)
- 3.44738 bar (50 psi)
- 0.5515808 bar (8 psi)
- 1.3100044 bar (19 psi)
- 2.3442184 bar (34 psi)
- 0.5515808 bar (8 psi)
- 1.65 bar (24 psi)
- 2.757904 bar (40 psi)
- 0.8963188 bar (13 psi)
- Max. temperature:
- 132 °C (270 °F)
- 132 °C (270 °F)
- 132 °C (270 °F)
- 132 °C (270 °F)
- 132 °C (270 °F)
- 132 °C (270 °F)
- 132 °C (270 °F)
- 132 °C (270 °F)
- 132 °C (270 °F)
- 132 °C (270 °F)
- 132 °C (270 °F)
- 132 °C (270 °F)
- 132 °C (270 °F)
- 132 °C (270 °F)
- 132 °C (270 °F)
- 132 °C (270 °F)
- Regulatory approvals:
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- Sterilisation method autoclave:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method EtO:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes