Masterflex® Transfer Tubing, Terminator®, Avantor®
Fournisseur: VWR International
Certificats
A propos de cet article
Superior abrasion resistance at high pressure.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Outstanding resistance to heat and ozone
Masterflex® Terminator® transfer tubing is ideal for use with severe service applications due to its superior abrasion resistance. The Buna-N nitrile tube, modified Buna-N nitrile tube cover, along with the high tensile textile core tube reinforcement makes this tubing suitable for high pressure applications. The tube and tube cover meet RMA Class A high oil resistance and nonconductive at 1000 VDC certification.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.
Spécifications
- Model:
- 95954-05
- 95954-03
- 95954-11
- 95954-07
- 95954-01
- Bend radius:
- 152.4 mm (6")
- 127 mm (5")
- 355.6 mm (14")
- 203.2 mm (8")
- 76.2 mm (3")
- Length per pack:
- 15.2 m (50 ft.)
- 30.5 m (100 ft.)
- 7.6 m (25 ft.)
- 15.2 m (50 ft.)
- 30.5 m (100 ft.)
- Max. pressure:
- 34.6 bar (501 psi)
- 34.6 bar (501 psi)
- 34.6 bar (501 psi)
- 34.6 bar (501 psi)
- 34.6 bar (501 psi)
- Max. temperature:
- 100 °C
- 100 °C
- 100 °C
- 100 °C
- 100 °C
- Sterilisation method autoclave:
- No
- No
- No
- No
- No
- Sterilisation method EtO:
- No
- No
- No
- No
- No
- Sterilisation method gamma:
- No
- No
- No
- No
- No