Masterflex® Hose Clamps, Acetal Copolymer, Avantor®
Supplier: VWR International
Certificates
About this item
Reusable white acetal copolymer clamps will not rust or corrode.
- Resists corrosion
Recommended for continuous use from ambient temperature to 200ºF (93ºC). Clamps are nonconducting and stable in a wide variety of chemicals.
Once set, clamps stay put. To use, just position clamp on hose or tubing bundle and press the jaws together with a pair of pliers.
Frequently Bought Together