J.T.Baker® PCR Plate 96 Wells for Automation
Fournisseur: Ritter, part of Avantor
Certificats
A propos de cet article
J.T.Baker® PCR plates are constructed with two plastic components. A polycarbonate ultra-rigid frame for automation and polypropylene ultra-thin wells for optimal thermal transfer and qPCR results.
- Available in barcoded and non barcoded version
- Standard SBS format for use with automation
- Slightly raised well rims for better sealing
- Stackable and comes with 8 notches for easy insertion on thermoblock
- White frame, white wells
- Alphanumeric well markings
- V-bottom
- 200 µl total volume
The wells are white to ensure high quality qPCR results by concentrating fluorescent signal, preventing loss of signal, and allowing for better signal detection by the instrument.
Applications: PCR, qPCR, NGS library set up.
Certifications: Produced under cleanroom conditions (ISO 14644-1 Class 8). Free from DNA, DNase, RNase, and PCR inhibitors.
Packaging: 10 packs of 5 plates in PE foil.
Spécifications
- Catalog No:
- 43001-0305
- 43001-0301
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)