High-Temperature Tube Furnaces with SiC Rod Heating up to 1500 °C, RHTC Series, Nabertherm
Fournisseur: NABERTHERM INC.
Certificats
A propos de cet article
The RHTC 80/230/15, RHTC 80/450/15, RHTC 80/710/15 high-temperature tube furnaces are heated by SiC heating elements which are arranged parallel to the working tube which provides excellent temperature uniformity.
- Tmax 1500 °C
- Represent an inexpensive variant in the high-temperature range
- SiC heating elements are easy to replace
- Type S thermocouple
- Active cooling of housing for low surface temperatures
- Programable: 5 programs, 4 segments
Working tube contains charge control with temperature measurement. Over-temperature limiter with adjustable cutout temperature is used to protect the furnace and load.
5 programs, 4 segments
Customers Who Bought This Also Bought