Vacuum Tubing, Flexible Stainless Steel, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
Flexible stainless steel accordion vacuum tubing featuring welded NW (DN) flanges.
- Welded NW flanges
- Glass to metal connection
Connect a NW glass flange to a vacuum pump.
Delivery: Includes silicone O-Rings.
Customers Who Bought This Also Bought