Metal to Metal Connection, NW Flange, Ace Glass
Supplier: Ace Glass
Certificates
About this item
Connections for NW metal flanges to like NW metal flanges.
- Connect vacuum pump to stainless steel tubing
- Various clamp and gasket options available
Included Silicone O-ring is suitable for use in temperatures from –60 to +205 °C. Viton O-ring is suitable for use in temperatures from –26 to +240 °C.
Delivery: Gasket and clamp sold separately.
Frequently Bought Together