D.T. Buddy Delivery Tube Hand Saver
# de catalogue 470337-874
Fournisseur: Shiv Dial Sud & Sons
Spécifications
- Description:D.T. Buddy (Delivery Tube Hand Saver)
- Size:130 mm
- Cat. no.:470337-874
Spécifications
A propos de cet article
The perfect hand-saving tool for inserting glass tubing into stoppers!
- Easy to use
- Made from good-quality hard rubber for easy gripping
- Protects hands by eliminating accidents while inserting glass tubing into stoppers
This tool prevents injury to hands due to shattered glass tubing. Simply push the tool through the stopper with a twist, remove the nose cone, insert the glass tube, and remove the stopper from the tool. The glass tubing remains in place in the stopper. Also useful for removing glass tubing from stoppers. Works on both rubber and cork stoppers.
Ordering information: Stopper and glass tubing not included.
Customers Who Bought This Also Bought