Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Spécifications
- Sterility:Sterile
- Description:AccuThrive #10S dermaplane blade with microcoat
- Material:Stainless steel
- Cat. no.:76539-030
- Packaging:500 blades/case
Spécifications
À propos de cet article
Coated #10S blade for dermaplaning.
- Coated with MicroCoat for sharpness and durability
AccuThrive's MicroCoat blade enrichment technology adds a layer of protection for each blade, helping maintain a sharp effective edge over multiple uses.