Accessories for Matrix Orbital Delta Plus and Matrix Delta Plus Thermoshakers
Fournisseur: IKA Works
Certificats
A propos de cet article
The ideal attachment design ensures an optimal temperature transfer and thus guarantees valid results. The thermoshakers Matrix Orbital Delta plus and Matrix Delta plus automatically recognize, which attachment is connected to the device via an interface in the locking mechanism. The attachment can be changed quickly and easily with an ergonomic lever.
- The heatable ThermoCover prevents condensation on the vial lid and wall. At the same time it enables constant reaction conditions
As part of IKA's Integrated Safety Concept, the insulating plastic coating of the attachment ensures user safety, as high temperatures are not transferred to the frame.
Customers Who Bought This Also Bought