Specimen Jars, Screw-Cap, Glass, United Scientific
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
These wide mouth jars are made from soda lime glass and have a polypropylene cap with a polyethylene liner.
- Autoclavable
Wide mouth jars feature a large opening, making it is easy to insert and remove materials.
Customers Who Bought This Also Bought