BRAND® Microcentrifuge Tubes with Attached Lids, 1.5 ml, BrandTech
Fournisseur: Brandtech
Certificats
A propos de cet article
These 1.5 ml tubes fit most common microcentrifuges.
- Withstands 20000 RCF
- Available in clear and five colors
- Non sterile or sterile
BRAND® microcentrifuge tubes fit most common rotors. These 1.5 ml tubes may be spun up to 20000 RCF (g) at 20 °C for 20 minutes. Manufactured from virgin polypropylene, these highly translucent tubes are free from flash, air bubbles and streaks. Flat caps form a tight, leak-free seal that is easy to open, even when wearing gloves.
Ordering information: Sold in bulk bags.
Customers Who Bought This Also Bought