Wind Up Tape Measure
# de catalogue 470337-816
Fournisseur: Wards
Spécifications
- Length:30 m (100 ft.)
- Description:Wind-up tape measure
- Cat. no.:470337-816
Spécifications
A propos de cet article
This flexible tape measure comes conveniently in a wind-up roll for easy use.
- Made of non-conducting material
- Waterproof, washable, and water-resistant
- Length of 30 meters
Tape is made of fiberglass with PVC covering. One side is in metric and the other in feet/inches. The case is made of a zinc-coated steel sheet, covered with vinyl and fitted with a flush winding-handle.
Customers Who Bought This Also Bought