R-MicronQuick® Microfiber Wipes, Vileda Professional®
Supplier: Vileda Professional - FHP
Certificates
About this item
Superior microfiber cleaning performance made from 70% recycled material.
- Superior cleaning performance
- Microfiber construction with up to 70% recycled material
- Streak-free cleaning, no water residue left on surface
- Lint-free
- Easy to wring and rinse
- Minimum 400 washings
- Available in 4 colors to differentiate wipes
Vileda Professional® believes every product developed must contribute to a sustainable future. That's resulted in the creation of a range of reuseable wipes that are manufactured with up to 70% recycled superior microfiber technology.
This is all part of a drive to help users reduce their footprint by reducing waste and CO₂ emissions throughout the cleaning process.
Specifications
- Catalog No:
- 76537-092
- 76537-094
- 76537-090
- Environmentally Preferable:
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials