A propos de cet article
Gravity convection provides a lower chance of sample cross contamination.
- PID technology for high precision control and small temperature fluctuation
- Extended-life silicone rubber gasket provides a long service life
- Features corrosion-resistant stainless steel
- Anti-skid shelf design, easy to operate by single hand
- Glass doors ensure perfect observation of the samples
Gravity convection incubators allow heat to fill the chamber naturally, providing a lower chance of cross contamination between samples. These gravity convection incubators have a temperature range from ambient +5 to 65 ℃. The inner chamber is constructed of stainless steel with a round, curved inner angle for easy cleaning. The anti-skid shelf design is easy to use with a single hand. Smaller models have a large outer glass door while larger models have an internal glass door. Glass doors avoid unnecessary heat dissipation and temperature changes when samples need to be viewed. The LCD display shows time, set temperature, actual temperature, and running status. Safety features include an auto start feature after power loss/return, temperature deviation alarm, over-temperature set point alarm, and over current protection.
Temperature resolution: 0.1 °C
Spécifications
- Catalog No:
- 76539-142
- 76539-144
- 76539-146
- 76539-148
- 76539-150
- Model:
- 10 L
- 35 L
- 55 L
- 115 L
- 210 L
- Convection type:
- Natural convection
- Natural convection
- Natural convection
- Natural convection
- Natural convection
- Int. W×D×H:
- 10.625×10.2×9.06"
- 14.375×12.6×11.8125"
- 15.75×12.9×14.9"
- 20.47×17.72×19.09"
- 25.6×19.6875×25.6"
- Ext. W×D×H:
- 20.08×15.75×13.8"
- 23.625×18.1×16.5"
- 26.1875×18.5×23.625"
- 30.9×23.2×28.16"
- 36.025×25.9×34.25"
- Temperature control:
- P.I.D. temperature controller
- P.I.D. temperature controller
- P.I.D. temperature controller
- P.I.D. temperature controller
- P.I.D. temperature controller
- Electrical:
- 120 VAC
- 120 VAC
- 120 VAC
- 120 VAC
- 120 VAC
- Includes:
- 1 chamber
- 1 chamber
- 1 chamber
- 1 chamber
- 1 chamber