Pipette/Instrument Organizer Tray, United Scientific
# de catalogue 470332-806
Fournisseur: United Scientific Supplies
Spécifications
- Description:Pipette/Instrument organizer tray
- Cat. no.:470332-806
Spécifications
A propos de cet article
Durable polypropylene tray for organizing pipettes or other instruments.
- Tray measures 16×12×1"
- Ideal for drawers, refrigerators or bench tops
- Tray walls are ¹/₈" thick
- Autoclavable
The pipette tray is designed to fit large drawers, refrigerators, and benchtops. Tray features 4 deep columns (approx. 0.75" deep) and is recommended for 1 to 10 ml pipettes. Molded-in polypropylene with ¹/₈" thick walls.
Customers Who Bought This Also Bought