Test Tubes, Polypropylene, United Scientific
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Round-bottom polypropylene test tubes.
- 12×75 mm tubes
- 5 ml capacity
- Autoclavable
Tubes are available in packs of 100 or 250.
Customers Who Bought This Also Bought