Bimetal Strip/Compound Bar, United Scientific
# de catalogue 470332-608
Fournisseur: United Scientific Supplies
Spécifications
- Description:Bimetal strip/Compound bar
- Cat. no.:470332-608
Spécifications
A propos de cet article
Simple tool to explore the thermal properties of matter.
- Bimetal strip is composed of two alloys that display different responses when exposed to heat
- Strips are securely bonded and fastened to a sturdy wooden handle
- Activity guide included
A quick and easy tool for demonstrating thermal properties of matter. Bar is bi-metallic and has a wooden handle attached. When heated, the bar bends dramatically.
Customers Who Bought This Also Bought