Updated Wimshurst Machine, United Scientific
# de catalogue 470332-762
Fournisseur: United Scientific Supplies
Spécifications
- Description:Updated Wimshurst machine
- Cat. no.:470332-762
Spécifications
A propos de cet article
A superior quality Wimshurst machine for demonstrating high voltage electric charges.
- Demonstrate the build up of charge
- Vividly demonstrate the strength of a stored charge and the distance for to jump to ground
This unit features acrylic rotating disks, good quality foil strips and brushes, insulated cylindrical capacitors, and a durable plastic platform. Guaranteed to produce large sparks for easy classroom demonstration.
Customers Who Bought This Also Bought