Earth Sun Planetarium, United Scientific
# de catalogue 470332-726
Fournisseur: United Scientific Supplies
Spécifications
- Description:Earth sun planetarium
- Cat. no.:470332-726
Spécifications
A propos de cet article
Demonstrate the rotational motion of Earth on its axis, its orbit around the Sun, and the revolution of the moon around Earth with this high-quality model.
- Battery operated sun
- Moon rotates around the Earth
- Illustrates the different seasons
As the Sun casts a beam of light onto Earth (via an embedded LED), users can easily manipulate the positions of each body to examine how their dynamics correspond to seasons, eclipses, and lunar phases.
Customers Who Bought This Also Bought