ContactUsLinkComponent
Wood Block, United Scientific
Wood Block, United Scientific
# de catalogue 470332-650
BLOCK WOOD 1in X 1in X 1in
Wood Block, United Scientific
# de catalogue 470332-650
Spécifications
  • Description:
    Wood block, 1×1×1"
  • Cat. no.:
    470332-650
Spécifications
A propos de cet article
Cube, 1" on each side. Made of pine.
  • Uniform sides
  • Dried out Pine wood
This pine block is perfect for investigating density or buoyancy.
Customers Who Bought This Also Bought