ContactUsLinkComponent
Advanced Projection Ripple Tank, United Scientific
Advanced Projection Ripple Tank, United Scientific
# de catalogue 470332-632
ADVANCED PROJECTION RIPPLE TANK
Advanced Projection Ripple Tank, United Scientific
# de catalogue 470332-632
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Advanced projection ripple tank
  • Cat. no.:
    470332-632
Spécifications
A propos de cet article
The Advanced Projection Ripple Tank is a comprehensive system for demonstrating basic wave phenomena using water waves.
  • Variable frequency generator
  • Large vertical display
  • Bright LED lights
A horizontal glass plate is mounted in a water-tight frame with foam beaches around the perimeter to suppress unwanted wave reflections. This tank is mounted on three tall legs with an angled mirror beneath the glass plate. A translucent screen is fitted to the front of the apparatus to display the projected image. A stray light shield can be placed around the back and sides of the apparatus to prevent ambient light from reaching the screen. In use, a pool of water a few millimeters deep is filled into the tank. A ripple generator for mounting and energizing ripplers is fitted to one side of the tank and an LED stroboscope lamp is adjustably mounted above the glass plate on a gooseneck support to provide the projection light. The ripple generator and the LED stroboscope lamp are both driven by a Wave Monitor control unit that synchronizes the wave generator’s oscillations with the stroboscope flashes to produce a stationary projected image. The wave generator can also be operated asynchronously, allowing the stroboscope flash frequency to be varied while the wave generator frequency remains fixed. This generates a moving projected image of variable speed. The included accessory set contains a series of five different ripplers (two of each), a pair of acrylic blocks, a barrier with slits, a clear foil with a printed grid, a plastic dropper, and a metric ruler.
Customers Who Bought This Also Bought