Coriolis Effect, United Scientific
Catalog # 470332-612
Supplier: United Scientific Supplies
Specifications
- Description:Coriolis effect
- Cat. no.:470332-612
Specifications
About this item
This apparatus helps students to understand the Coriolis effect, the phenomenon whereby an object moving on a spinning surface will travel in a curved path.
- Components include:
- Turntable
- Erasable tracing surface
- Ramp
- Steel balls (2)
The demonstrations allow students to observe the Coriolis effect on a small scale, then explore how the Earth’s constant rotation influences wind patterns and ocean currents around the world.
Customers Who Bought This Also Bought