Flask Form Electroscope, United Scientific
# de catalogue 470325-664
Fournisseur: United Scientific Supplies
Spécifications
- Description:Flask form electroscope
- Cat. no.:470325-664
Spécifications
A propos de cet article
Classroom electroscope housed in a 250 ml borosilicate flask.
- Activity guide included
This electroscope is housed in a 250 ml borosilicate flask with a metal rod attached to a rubber stopper. A pair of aluminum leaves is suspended from an alligator clip. The knob on top of the rubber stopper accepts a charge. When the charge enters the flask, the aluminum leaves separate.
Customers Who Bought This Also Bought