ContactUsLinkComponent
Flask Form Electroscope, United Scientific
Flask Form Electroscope, United Scientific
# de catalogue 470325-664
Flask Form Electroscope, housed in a 250ml borosilicate flask with a metal rod attached to a rubber stopper. A pair of aluminum leaves is suspended from an alligator clip. The knob on top of the rubber stopper accepts a charge. Includes activity guide.
Flask Form Electroscope, United Scientific
# de catalogue 470325-664
Spécifications
  • Description:
    Flask form electroscope
  • Cat. no.:
    470325-664
Spécifications
A propos de cet article
Classroom electroscope housed in a 250 ml borosilicate flask.
  • Activity guide included
This electroscope is housed in a 250 ml borosilicate flask with a metal rod attached to a rubber stopper. A pair of aluminum leaves is suspended from an alligator clip. The knob on top of the rubber stopper accepts a charge. When the charge enters the flask, the aluminum leaves separate.
Customers Who Bought This Also Bought