Salus HC PAPR Kits, Bullard
Fournisseur: Bullard
Certificats
A propos de cet article
Ideal for healthcare and pharmaceutical applications.
- Unique shoulder carriage design for maximum comfort
- Auto dampening technology to minimize noise
- Closed cell foam; easy to clean
Unit features one-button operation located on chest, dual alarms, and two-speed blower. Comfortable hood cosntructed of Tychem 2000. Small, lightweight battery can easily be changed while operating the blower unit.
Certifications: NIOSH approved to P100 filter standard.
Customers Who Bought This Also Bought