A propos de cet article
Desktop winterization kit contains materials needed to efficiently separate compounds and refine oils.
- Large sized Buchner funnel
- Food grade PVC tubing
Each convenient package includes a table type buchner funnel, neoprene stopper, medium flow filter paper, food grade tubing, radiator clamps and a coupler. The tubing has a temperature range of −40 to 125 °F and it meets FDA standards and NSF 51. Our Desktop Winterization Labware Kit is available in three options: without glassware, with a 5000 ml filtering flask or a 10000 ml filtering bottle. The filtering flask is from our FG5340 series, which is constructed using heavy wall borosilicate and include a thick rim and hose barb. The 10000 ml set includes our FFB5340 series heavy wall borosilicate filtering bottle with hose barb. The porcelain table type buchner funnel is from our JTTBF series. Filter paper is from our FPR series and has medium porosity and flow rate. Circular filters are packaged 100 per box.
DWNTK Desktop Winterization Labware Kit Contents:
- (1) Buchner Funnel, Table Type, Porcelain, 1900 ml (JTTBF1900)
- (5) Food Grading PVC Tubing, 60" long, 1" Int.Ø×1¹/₄" Int.Ø×¹/₈" Wall
- (1) Filter Paper, Medium Porosity/Flow, 18 cm circle, pk/100 (FPR180)
- (2) Radiator Clamp
- (1) Neoprene Stopper, #13, 1" hole
- (1) Coupler, ³/₄" Int.Ø
DWNTK5 5000 ml Winterization Labware Kit Contents:
- (1) Filtering Flask, Borosilicate Glass, 5000 ml (FG5340-5000)
- (1) Buchner Funnel, Table Type, Porcelain, 1900 ml (JTTBF1900)
- (5) Food Grading PVC Tubing, 60” long, 1" Int.Ø×1¹/₄" Int.Ø×¹/₈" Wall
- (1) Filter Paper, Medium Porosity/Flow, 18 cm circle, pk/100 (FPR180)
- (2) Radiator Clamp
- (1) Neoprene Stopper, #13, 1" hole
- (1) Coupler, ³/₄" Int.Ø
DWNTK10 10000 ml Winterization Labware Kit Contents:
- (1) Filtering Bottle, Borosilicate Glass, 10000 ml (FFB5340-10000)
- (1) Buchner Funnel, Table Type, Porcelain, 1900 ml (JTTBF1900)
- (5) Food Grading PVC Tubing, 60" long, 1" Int.Ø×1¹/₄" Int.Ø×¹/₈" Wall
- (1) Filter Paper, Medium Porosity/Flow, 18 cm circle, pk/100 (FPR180)
- (2) Radiator Clamp
- (1) Neoprene Stopper, #13, 1" hole
- (1) Coupler, ³/₄" Int.Ø