Graduated Cylinders, Class B, Borosilicate Glass, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Fine quality borosilicate glass cylinders .
- Features double metric scale
- Stable hexagon base
Double metric scale, calibrated to contain. Borosilicate glass cylinders with pour spouts feature heavy uniform wall tubing and attached glass hexagonal bases. All cylinders come with bumper guards.
Certifications: Cylinders comply with ASTM E1272, Class B standards.
Customers Who Bought This Also Bought