Disposable Culture Tubes, Borosilicate, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Disposable culture tubes manufactured from high-quality borosilicate glass, with rimless design for easy plugging using a stopper.
- Rimless design for easy plugging with stopper (not included)
- Available in multiple sizes
Tubes meet ASTM standards and are alkali-free and inert.
Customers Who Bought This Also Bought