Specific Gravity Bottles, Borosilicate Glass, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Borosilicate glass bottle used to determine specific gravity.
- Uncalibrated
- Autoclavable
Includes a PTFE stopper with a hole to allow air to escape.
Customers Who Bought This Also Bought