Pipettes, Serological, Accuracy as per Class B, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Type 3 serological pipettes with color-coding.
- Printed with durable amber markings at intervals
- Color-coded in white for easy identification of capacity
Made from borosilicate glass in compliance with USP, Class B standards. Calibrated to deliver.
Customers Who Bought This Also Bought