Pipettes, Measuring (Mohr), Class B, United Scientific Supplies
Supplier: United Scientific Supplies
Certificates
About this item
Color-coded Mohr pipettes.
- Printed with durable amber markings
- Color-coded in white for easy identification of capacity
Made from borosilicate glass in compliance with ASTM specification E1293 for Class B pipettes. Calibrated to deliver.
Customers Who Bought This Also Bought