Pipettes, Measuring (Mohr), Class A, Batch Certified, United Scientific Supplies
Color-coded Mohr pipettes.
- Printed with durable amber markings
- Color-coded in white for easy identification of capacity
Made from borosilicate glass in compliance with ASTM specification E1293. Batch certified and calibrated to deliver.
Customers Who Bought This Also Bought