Pipettes, Measuring (Mohr), Class A, Individually Certified, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Color-coded Mohr pipettes.
- Printed with durable amber markings
- Color-coded in white for easy identification of capacity
Made from borosilicate glass in compliance with ASTM specification E1293. Individually serialized and certified; calibrated to deliver.
Customers Who Bought This Also Bought