Distilling Flask, Borosilicate Glass, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Flasks with a long side arm for distillation applications.
- Features large white block lettering for easy identification of capacity
Made from low-expansion borosilicate glass, these flasks feature uniform wall thickness for superior mechanical strength and shock resistance.
Customers Who Bought This Also Bought