Graduated Cylinders, Glass, Class A, Individually Certified, with Stopper, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Borosilicate glass cylinders feature heavy uniform wall tubing and strong, stable, hexagonal bases. All cylinders come with bumper guards.
- With hollow glass stopper
- Double metric scale, calibrated to contain
Cylinders comply with ASTM E1272, Class A, meets USP standards, individually serialized and certified.
Customers Who Bought This Also Bought