Burette, Class B, PTFE Stopcock, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Made from high-quality accurate bore tubing for a high degree of accuracy, reliability and durability.
- Amber graduations at intervals
- Available in multiple sizes
Includes PTFE straight bore stopcock.
Customers Who Bought This Also Bought