Burette, Class A, PTFE Stopcock, Batch Certified, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Class A burette, per ASTM E287. Meets USP standards and is batch certified.
- Graduated at intervals with a waiting time of 30 seconds
- Includes PTFE straight bore stopcock
- Available in multiple sizes
Manufactured from precision bore tubing, calibrated to deliver (TD, Ex) on computer-controlled machines.
Customers Who Bought This Also Bought